The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001110209.1 | NP_001103679.1 | 13 - 14 | 1312 | 116 |
GenBank mRNA | AK014897.1 | - | 13 - 14 | 1209 | 116 |
AK137445.1 | - | 13 - 14 | 1312 | 116 |