The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_016891.3 | NP_058587.1 | 7 - 8 | 967 | 90 |
GenBank mRNA | AB021743.1 | - | 7 - 8 | 915 | 90 |
AK054239.1 | - | 7 - 8 | 927 | 90 | |
AK078135.1 | - | 7 - 8 | 946 | 90 | |
AK088776.1 | - | 7 - 8 | 939 | 90 | |
BC006606.1 | - | 7 - 8 | 967 | 90 | |
BC052678.1 | - | 7 - 8 | 963 | 90 |