The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_029104.1 | NP_083380.1 | 3 - 4 | 668 | 65 |
GenBank mRNA | AK014794.1 | - | 3 - 4 | 449 | 65 |
BC119773.1 | - | 3 - 4 | 561 | 65 | |
BC119816.1 | - | 3 - 4 | 561 | 65 |