Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Ppp2ca
Entrez Gene ID
19052
Gene Name
protein phosphatase 2 (formerly 2A), catalytic subunit, alpha isoform
Gene Aliases
PP2A, R75353
Chromosome Location
Chr.11: 52098824 - 52122749 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_019411.4 NP_062284.1 6 - 7 1081 74
GenBank mRNA AF076192.1 - 6 - 7 1051 74
AK076110.1 - 6 - 7 1082 74
AK172644.1 - 6 - 7 1054 74
BC003856.1 - 6 - 7 1058 74
BC054458.1 - 6 - 7 1041 74
Z67745.1 - 6 - 7 856 74

View Full Product Details