Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Ankrd34b
Entrez Gene ID
218440
Gene Name
ankyrin repeat domain 34B
Gene Aliases
6430502M16Rik, DP58
Chromosome Location
Chr.13: 92425641 - 92441658 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_175455.4 NP_780664.2 3 - 4 462 91
GenBank mRNA AK078203.1 - 3 - 4 536 91
AK090387.1 - 3 - 4 463 91
AK138465.1 - 2 - 3 648 91
BC096609.1 - 3 - 4 338 91
BC119312.1 - 3 - 4 324 91
BC119314.1 - 3 - 4 324 91

View Full Product Details