Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Axdnd1
Entrez Gene ID
77352
Gene Name
axonemal dynein light chain domain containing 1
Gene Aliases
9430070O13Rik, Gm979
Chromosome Location
Chr.1: 156330415 - 156420915 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
GenBank mRNA AK134071.1 - 6 - 7 665 88
BC150932.1 - 5 - 6 517 88
BC172088.1 - 5 - 6 607 88

View Full Product Details