Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Tbx4
Entrez Gene ID
21387
Gene Name
T-box 4
Gene Aliases
3930401C23
Chromosome Location
Chr.11: 85886343 - 85916099 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_011536.2 NP_035666.2 7 - 8 1049 83
NM_172798.1 NP_766386.1 6 - 7 999 83
GenBank mRNA AB221665.1 - 7 - 8 1049 83
AK028407.1 - 6 - 7 933 83
AK147984.1 - 6 - 7 917 83
AY075134.1 - 7 - 8 1049 83

View Full Product Details