The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_026684.2 | NP_080960.1 | 1 - 2 | 202 | 83 |
GenBank mRNA | AK002517.1 | - | 1 - 2 | 166 | 83 |
BC031664.1 | - | 1 - 2 | 202 | 83 | |
BC043013.1 | - | 1 - 2 | 179 | 83 |