Assay Design
The assay primers and probes lie within a single exon. This assay will detect gDNA.
Gene Transcripts
- Gene Symbol
- Arxes1
- Entrez Gene ID
-
76219
- Gene Name
- adipocyte-related X-chromosome expressed sequence 1
- Gene Aliases
-
6530401D17Rik, Spc22/23, Spcs3
- Chromosome Location
-
Chr.X: 136033367 - 136034946 on Build GRCm38
Non Gene Transcripts
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
IMAGE Clone ID |
Transcript Location |
Amplicon Length |
GenBankmRNA |
CJ294638.1 |
- |
1 |
411 |
|
Chr. X - 136034438 - 136034942 [+] on Build GRCm38 |
126 |
GenBankmRNA |
AI839310.1 |
- |
2 |
|
|
Chr. X - 136034542 - 136038447 [-] on Build GRCm38 |
125 |
GenBankmRNA |
BU700875.1 |
- |
1 |
|
|
Chr. X - 136034427 - 136034943 [-] on Build GRCm38 |
126 |
GenBankmRNA |
CJ237971.1 |
- |
1 |
374 |
|
Chr. X - 136034469 - 136034940 [+] on Build GRCm38 |
126 |
GenBankmRNA |
BB041721.2 |
- |
1 |
579 |
|
Chr. X - 136034266 - 136034925 [+] on Build GRCm38 |
126 |
Adjacent Gene Details
|
Gene Name |
Gene Symbol |
Alias |
Distance |
Relative Direction |
Location Chromosome |
Upstream |
adipocyte-related X-chromosome expressed sequence 2 |
Arxes2 |
2900062L11Rik, Spc22/23, Spcs3 |
-38.0kb
|
Sense |
Chr. - 135993820 - 135995359 on Build GRCm38 |
Downstream |
brain expressed X-linked 2 |
Bex2 |
AL024066, Bex1 |
31.6kb
|
Anti-sense |
Chr. - 136066565 - 136068236 on Build GRCm38 |
View Full Product Details