The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NR_104310.1 | 2 - 3 | 1770 | 71 | |
GenBank mRNA | U95145.1 | - | 2 - 3 | 1774 | 71 |