Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Pcnx
Entrez Gene ID
54604
Gene Name
pecanex homolog (Drosophila)
Gene Aliases
2900024E21Rik, 3526401J03Rik, AF096286, AI327143, AI413187, Pcnxl1
Chromosome Location
Chr.12: 81860028 - 82000924 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_018814.3 NP_061284.2 22 - 23 4688 73
GenBank mRNA AK129221.1 - 17 - 18 2568 73
AK154122.1 - 3 - 4 416 73
AK170320.1 - 3 - 4 416 73
BC057333.1 - 15 - 16 2431 73
BC066849.1 - 17 - 18 2311 73
BC098370.1 - 17 - 18 2278 73

View Full Product Details