Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Pcnx
Entrez Gene ID
54604
Gene Name
pecanex homolog (Drosophila)
Gene Aliases
2900024E21Rik, 3526401J03Rik, AF096286, AI327143, AI413187, Pcnxl1
Chromosome Location
Chr.12: 81860028 - 82000924 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_018814.3 NP_061284.2 27 - 28 5457 80
GenBank mRNA AF096286.1 - 21 - 22 3287 80
AK014387.1 - 1 - 2 145 80
AK129221.1 - 22 - 23 3337 80
AK154122.1 - 8 - 9 1185 80
AK170320.1 - 8 - 9 1185 80
BC057333.1 - 20 - 21 3200 80
BC098370.1 - 22 - 23 3047 80

View Full Product Details