Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Pcnx
Entrez Gene ID
54604
Gene Name
pecanex homolog (Drosophila)
Gene Aliases
2900024E21Rik, 3526401J03Rik, AF096286, AI327143, AI413187, Pcnxl1
Chromosome Location
Chr.12: 81860028 - 82000924 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_018814.3 NP_061284.2 33 - 34 6657 86
GenBank mRNA AF096286.1 - 27 - 28 4487 86
AK014387.1 - 7 - 8 1345 86
AK129221.1 - 28 - 29 4537 86
AK154122.1 - 14 - 15 2385 86
AK170320.1 - 14 - 15 2385 86
BC066849.1 - 28 - 29 4322 86
BC098370.1 - 28 - 29 4247 86

View Full Product Details