Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
Cwc27
Entrez Gene ID
67285
Gene Name
CWC27 spliceosome-associated protein homolog (S. cerevisiae)
Gene Aliases
3110009E13Rik, NY-CO-10, Sdccag10
Chromosome Location
Chr.13: 104631322 - 104816964 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_026072.1 NP_080348.1 5 - 6 718 142
GenBank mRNA AK014025.1 - 5 - 6 701 142
AK075902.1 - 5 - 6 696 142
AK077779.1 - 5 - 6 661 142
AK166770.1 - 5 - 6 718 142
BC025437.1 - 5 - 6 518 142

View Full Product Details