The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_011934.4 | NP_036064.3 | 1 - 2 | 323 | 87 |
GenBank mRNA | AK032025.1 | - | 1 - 2 | 131 | 87 |
AK044339.1 | - | 1 - 2 | 324 | 87 | |
BB642270.1 | - | 1 - 2 | 324 | 87 |