The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001080943.2 | NP_001074412.1 | 1 - 2 | 533 | 72 |
GenBank mRNA | BC128021.1 | - | 1 - 2 | 537 | 72 |