Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Pcnx
Entrez Gene ID
54604
Gene Name
pecanex homolog (Drosophila)
Gene Aliases
2900024E21Rik, 3526401J03Rik, AF096286, AI327143, AI413187, Pcnxl1
Chromosome Location
Chr.12: 81860028 - 82000924 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_018814.3 NP_061284.2 2 - 3 704 82
GenBank mRNA CF746564.1 - 1 - 2 127 82
CJ107156.1 - 2 - 3 371 82

View Full Product Details