The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001029990.1 | NP_001025161.1 | 3 - 4 | 475 | 73 |
GenBank mRNA | AK009576.1 | - | 3 - 4 | 389 | 73 |
AK032357.1 | - | - | 466 | 73 | |
AK087101.1 | - | 3 - 4 | 407 | 73 | |
AK155096.1 | - | 3 - 4 | 394 | 73 | |
AK156895.1 | - | 2 - 3 | 525 | 73 | |
AK172330.1 | - | 2 - 3 | 525 | 73 | |
BC138944.1 | - | 3 - 4 | 428 | 73 | |
BC145472.1 | - | 3 - 4 | 428 | 73 | |
BC145473.1 | - | 2 - 3 | 511 | 73 | |
BC145474.1 | - | 3 - 4 | 428 | 73 | |
CF172670.1 | - | 3 - 4 | 475 | 73 |