The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_028626.1 | NP_082902.1 | 2 - 3 | 418 | 65 |
GenBank mRNA | AK003511.1 | - | 2 - 3 | 415 | 65 |
BC038157.1 | - | 2 - 3 | 418 | 65 |