The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001291184.1 | NP_001278113.1 | 20 - 21 | 2780 | 65 |
NM_011048.1 | NP_035178.1 | 21 - 22 | 2819 | 65 | |
GenBank mRNA | AF008222.1 | - | 8 - 9 | 795 | 65 |
AK081039.1 | - | 3 - 4 | 337 | 65 | |
BC003302.1 | - | 5 - 6 | 546 | 65 | |
BC025946.1 | - | 5 - 6 | 550 | 65 | |
BC037450.1 | - | 18 - 19 | 2385 | 65 | |
D50060.1 | - | 20 - 21 | 2703 | 65 |