The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001278219.1 | NP_001265148.1 | 1 - 2 | 204 | 56 |
NM_026866.3 | NP_081142.3 | 1 - 2 | 204 | 56 | |
GenBank mRNA | AK138276.1 | - | 1 - 2 | 161 | 56 |
AK170113.1 | - | 1 - 2 | 199 | 56 | |
BQ964652.1 | - | - | 209 | 56 |