Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Cxxc4
Entrez Gene ID
319478
Gene Name
CXXC finger 4
Gene Aliases
9330210J02Rik, C030003J12Rik, Idax
Chromosome Location
Chr.3: 134236415 - 134262090 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001004367.4 NP_001004367.2 3 - 4 1584 107
GenBank mRNA BC065149.1 - 4 - 5 1270 107
BC067052.1 - 3 - 4 1114 107
BC082332.1 - 2 - 3 801 107

View Full Product Details