Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
Vmn2r65
Entrez Gene ID
100009609
Gene Name
vomeronasal 2, receptor 65
Gene Aliases
ENSMUSG00000070600, V2ro
Chromosome Location
Chr.7: 84940169 - 84964009 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001105180.1 NP_001098650.1 4 - 5 1483 147
GenBank mRNA AB194095.1 - 2 - 3 536 147

View Full Product Details