The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_008102.3 | NP_032128.1 | 4 - 5 | 656 | 85 |
GenBank mRNA | AK150657.1 | - | 4 - 5 | 668 | 85 |
AK152571.1 | - | 4 - 5 | 668 | 85 | |
BC005643.1 | - | 4 - 5 | 656 | 85 | |
BC069921.1 | - | 4 - 5 | 660 | 85 | |
L09737.1 | - | 4 - 5 | 643 | 85 |