Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Bbs5
Entrez Gene ID
72569
Gene Name
Bardet-Biedl syndrome 5 (human)
Gene Aliases
1700049I01Rik, 2700023J09Rik
Chromosome Location
Chr.2: 69646730 - 69667569 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_028284.2 NP_082560.1 9 - 10 852 127
GenBank mRNA AK006743.1 - 8 - 9 809 127
AK012283.1 - 9 - 10 852 127
BC061031.1 - 9 - 10 853 127

View Full Product Details