The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_020575.2 | NP_065600.1 | 9 - 10 | 2228 | 85 |
GenBank mRNA | AF155739.1 | - | 9 - 10 | 2228 | 85 |
AK143177.1 | - | 9 - 10 | 2250 | 85 | |
AK166666.1 | - | 9 - 10 | 2235 | 85 | |
AK167246.1 | - | 9 - 10 | 2210 | 85 | |
AK168555.1 | - | 9 - 10 | 2231 | 85 | |
BC025029.1 | - | 9 - 10 | 2192 | 85 |