Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
Arhgap1
Entrez Gene ID
228359
Gene Name
Rho GTPase activating protein 1
Gene Aliases
B230365D05Rik, C76222, Cdc42GAP, p50rhoGAP
Chromosome Location
Chr.2: 91650011 - 91672325 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001145902.1 NP_001139374.1 7 - 8 770 76
NM_146124.4 NP_666236.3 7 - 8 979 76
NR_027371.1 8 - 9 845 76
NR_027372.1 8 - 9 830 76
NR_027373.1 8 - 9 860 76
GenBank mRNA AK046291.1 - 7 - 8 770 76
AK079048.1 - 8 - 9 754 76
AK084622.1 - 7 - 8 980 76
AK085554.1 - 8 - 9 734 76
BC006592.1 - 8 - 9 753 76
BC089306.1 - 7 - 8 684 76

View Full Product Details