Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Klc1
Entrez Gene ID
16593
Gene Name
kinesin light chain 1
Gene Aliases
AI874768, Kns2
Chromosome Location
Chr.12: 111758849 - 111807844 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001025362.2 NP_001020533.2 14 - 15 1868 60

View Full Product Details