Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Pygb
Entrez Gene ID
110078
Gene Name
brain glycogen phosphorylase
Gene Aliases
-
Chromosome Location
Chr.2: 150786796 - 150831748 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_153781.1 NP_722476.1 16 - 17 2038 78
GenBank mRNA AK032769.1 - 16 - 17 2051 78
AK035673.1 - 16 - 17 1768 78
AK133794.1 - 7 - 8 768 78
AK134667.1 - 16 - 17 2048 78
AK142716.1 - 16 - 17 2048 78
AK147763.1 - 16 - 17 2070 78
AK168381.1 - 16 - 17 2050 78
AK169052.1 - 16 - 17 2073 78
AK169631.1 - 16 - 17 2052 78
BC031457.1 - 16 - 17 2045 78
BC032209.1 - 10 - 11 1193 78
BC035283.1 - 16 - 17 2038 78

View Full Product Details