Assay Design
The assay probe spans an exon junction.
Gene Transcripts
- Gene Symbol
- Ppapdc3
- Entrez Gene ID
-
227721
- Gene Name
- phosphatidic acid phosphatase type 2 domain containing 3
- Gene Aliases
-
D830019K17Rik, NET39
- Chromosome Location
-
Chr.2: 32095516 - 32110820 on Build GRCm38
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
Amplicon Length |
GenBank mRNA |
AK048016.1 |
- |
1 - 2 |
105 |
60 |
View Full Product Details