Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Elf5
Entrez Gene ID
13711
Gene Name
E74-like factor 5
Gene Aliases
ESE-2, ESE-5, ESE-5.
Chromosome Location
Chr.2: 103411693 - 103450989 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001145813.1 NP_001139285.1 1 - 2 27 65
GenBank mRNA AK132484.1 - 1 - 2 438 65

View Full Product Details