Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
Accs
Entrez Gene ID
329470
Gene Name
1-aminocyclopropane-1-carboxylate synthase (non-functional)
Gene Aliases
2610203E10Rik, Phacs
Chromosome Location
Chr.2: 93833467 - 93849995 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001290782.1 NP_001277711.1 6 - 7 742 74
NM_183220.3 NP_899043.1 6 - 7 797 74
GenBank mRNA AK011867.1 - 6 - 7 646 74
BC039569.1 - 6 - 7 774 74

View Full Product Details