Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
Ccdc22
Entrez Gene ID
54638
Gene Name
coiled-coil domain containing 22
Gene Aliases
AI481216, DXImx40e, Ppp1r3f, Sfc22
Chromosome Location
Chr.X: 7593809 - 7605420 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_138603.3 NP_613069.3 13 - 14 1672 67
GenBank mRNA AF229637.1 - 13 - 14 1613 67
AK010536.1 - 13 - 14 1590 67
AK039573.1 - 13 - 14 1645 67
BC011195.1 - 13 - 14 1577 67
BC057686.1 - 4 - 5 1722 67

View Full Product Details