Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
Zmat1
Entrez Gene ID
215693
Gene Name
zinc finger, matrin type 1
Gene Aliases
A230096A06, AW990744, B930008K04Rik
Chromosome Location
Chr.X: 134971372 - 135009209 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_175446.3 NP_780655.2 3 - 4 505 99
GenBank mRNA AK133258.1 - - 457 99
AW044958.1 - 3 - 4 507 99
BC067022.1 - 3 - 4 481 99

View Full Product Details