The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001031664.1 | NP_001026834.1 | 1 - 2 | 653 | 126 |
GenBank mRNA | BC055771.1 | - | 1 - 2 | 610 | 126 |
CX209897.1 | - | 1 - 2 | 653 | 125 |