Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Cep44
Entrez Gene ID
382010
Gene Name
centrosomal protein 44
Gene Aliases
4933440G23Rik, mKIAA1712
Chromosome Location
Chr.8: 56531516 - 56550777 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001009951.1 NP_001009951.1 2 - 3 155 65
GenBank mRNA AK220415.1 - 1 - 2 111 65
BC088983.1 - 2 - 3 155 65

View Full Product Details