Assay Design
The assay primers and probes lie within a single exon. This assay will detect gDNA.
Gene Transcripts
- Gene Symbol
- Maf
- Entrez Gene ID
-
17132
- Gene Name
- avian musculoaponeurotic fibrosarcoma (v-maf) AS42 oncogene homolog
- Gene Aliases
-
2810401A20Rik, A230108G15Rik, AW047063, c-maf
- Chromosome Location
-
Chr.8: 115703253 - 115706894 on Build GRCm38
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
Amplicon Length |
GenBank mRNA |
AK033026.1 |
- |
- |
2268 |
79 |
View Full Product Details