Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Zc3h3
Entrez Gene ID
223642
Gene Name
zinc finger CCCH type containing 3
Gene Aliases
BC049953, Kiaa0150, Smicl, Zc3hdc3
Chromosome Location
Chr.15: 75754434 - 75841971 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_172121.1 NP_742119.1 4 - 5 1717 84
GenBank mRNA AJ516034.1 - 4 - 5 1717 84
AK086799.1 - 2 - 3 214 84
AK138281.1 - 4 - 5 1725 84
BC038813.1 - 3 - 4 654 84
BC049953.1 - 3 - 4 573 84
BC060682.1 - 4 - 5 1716 84

View Full Product Details