Assay Design

The assay primers and probes lie within a single exon. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
Gpatch3
Entrez Gene ID
242691
Gene Name
G patch domain containing 3
Gene Aliases
D930035B09Rik, Gpatc3
Chromosome Location
Chr.4: 133574745 - 133584242 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
GenBank mRNA BC096468.1 - 7 - 7 1662 80

View Full Product Details