Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Mrgprb2
Entrez Gene ID
243979
Gene Name
MAS-related GPR, member B2
Gene Aliases
4833406I20Rik, Mgrg14
Chromosome Location
Chr.7: 48550965 - 48658094 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_175531.4 NP_780740.2 1 - 2 66 89
GenBank mRNA AK029369.1 - 1 - 2 67 89
BC107390.1 - 1 - 2 64 89
BC107391.2 - 1 - 2 64 89

View Full Product Details