Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Mrgpra1
Entrez Gene ID
233221
Gene Name
MAS-related GPR, member A1
Gene Aliases
MrgA1
Chromosome Location
Chr.7: 47334875 - 47354240 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_153095.2 NP_694735.2 1 - 2 49 143
GenBank mRNA AY042191.1 - 1 - 2 49 143

View Full Product Details