The assay primers and probes lie within a single exon. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_011221.3 | NP_035351.1 | 1 - 1 | 8378 | 115 |
GenBank mRNA | AK009365.1 | - | 1 - 1 | 1440 | 115 |
BC090992.1 | - | 1 - 1 | 1671 | 115 |