The assay primers and probes lie within a single exon. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_027149.2 | NP_081425.2 | 3 - 3 | 1801 | 88 |
GenBank mRNA | AK052481.1 | - | 3 - 3 | 1748 | 88 |
AK136417.1 | - | 3 - 3 | 1761 | 88 | |
AK145389.1 | - | 3 - 3 | 1724 | 88 | |
BC026761.1 | - | 4 - 4 | 1651 | 88 | |
BC064773.1 | - | 3 - 3 | 1758 | 88 |