Assay Design

The assay primers and probes lie within a single exon. This assay will detect gDNA.

Gene Transcripts

A Gene Symbol is not available because the non-coding assay was designed to a GenBank RNA or EST accession that does not map to an Entrez Gene ID.



Non Gene Transcripts

Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Transcript Location Amplicon Length
GenBankmRNA BB216870.2 - 1 458 Chr. 16 - 19212660 - 19213291 [-] on Build GRCm38 157
GenBankmRNA AK079984.1 - 1053 Chr. - - [-] on Build 157


Adjacent Gene Details

Gene Name Gene Symbol Alias Distance Relative Direction Location Chromosome
Upstream immunoglobulin lambda variable 1 Iglv1 1810027O01Rik, Igl-V1 -127.2kb Sense Chr. - 19085017 - 19085460 on Build GRCm38
Downstream immunoglobulin lambda variable 2 Iglv2 Igl-V2 47.1kb Sense Chr. - 19260403 - 19260844 on Build GRCm38

View Full Product Details