The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_175638.3 | NP_783569.1 | 18 - 19 | 3772 | 151 |
GenBank mRNA | AK008047.1 | - | 2 - 3 | 197 | 151 |
AY184228.1 | - | 18 - 19 | 3741 | 151 | |
BC026591.1 | - | 9 - 10 | 1711 | 151 | |
BC026679.1 | - | 11 - 12 | 1971 | 151 | |
BC037114.1 | - | 16 - 17 | 3096 | 151 | |
BC043677.1 | - | 18 - 19 | 3145 | 151 | |
BC096453.1 | - | 18 - 19 | 3728 | 151 |