The assay primers and probes lie within a single exon. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_026943.1 | NP_081219.1 | 3 - 3 | 372 | 85 |
GenBank mRNA | AK007389.1 | - | 3 - 3 | 351 | 85 |
AK045968.1 | - | 3 - 3 | 347 | 85 | |
AK088105.1 | - | 3 - 3 | 351 | 85 | |
BC043014.1 | - | 3 - 3 | 372 | 85 | |
BC051208.1 | - | 2 - 2 | 351 | 85 | |
BC116858.1 | - | 3 - 3 | 355 | 85 | |
BC116860.1 | - | 3 - 3 | 355 | 85 |