The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001033962.1 | NP_001029134.1 | 2 - 3 | 568 | 99 |
NM_011668.2 | NP_035798.2 | 4 - 5 | 755 | 99 | |
NM_173010.3 | NP_766598.1 | 2 - 3 | 570 | 99 | |
GenBank mRNA | AK029133.1 | - | 2 - 3 | 571 | 99 |
AK083795.1 | - | 2 - 3 | 499 | 99 | |
AK220549.1 | - | 2 - 3 | 468 | 99 | |
BC131658.1 | - | 2 - 3 | 556 | 99 | |
BC131659.1 | - | 2 - 3 | 556 | 99 | |
CD352481.1 | - | 4 - 5 | 652 | 99 | |
U96636.1 | - | 4 - 5 | 302 | 99 |