Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Kncn
Entrez Gene ID
654462
Gene Name
kinocilin
Gene Aliases
Kino, L5
Chromosome Location
Chr.4: 115884400 - 115887964 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001039124.3 NP_001034213.1 1 - 2 488 95
GenBank mRNA BC132034.1 - 1 - 2 435 95
BC132036.1 - 1 - 2 435 95
BC145375.1 - 1 - 2 435 95
DQ235452.1 - 1 - 2 488 95

View Full Product Details