Assay Design

The assay primers and probes lie within a single exon. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
Oog2
Entrez Gene ID
381570
Gene Name
oogenesin 2
Gene Aliases
C85069
Chromosome Location
Chr.4: 144190717 - 144196934 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_198661.3 NP_941063.1 5 - 5 1875 76
GenBank mRNA AY573562.1 - 5 - 5 1875 76
BC052839.1 - 5 - 5 1875 76

View Full Product Details