The assay primers and probes lie within a single exon. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001018087.1 | NP_001018097.1 | 1 - 1 | 230 | 150 |
GenBank mRNA | AK137057.1 | - | 1 - 1 | 340 | 150 |
BC052689.1 | - | 1 - 1 | 230 | 150 |